missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APH1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | APH1A |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 4-6ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18112484
|
Novus Biologicals
NBP2-27176SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18140554
|
Novus Biologicals
NBP2-27176 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APH1A Polyclonal specifically detects APH1A in Human, Bovine, Canine, Equine, Opossum, Primate samples. It is validated for Western Blot.Specifications
| APH1A | |
| Western Blot 4-6ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Bovine, Canine, Equine, Opossum, Primate | |
| 6530402N02Rik, anterior pharynx defective 1 homolog A (C. elegans), APH-1, APH-1A, aph-1alpha, CGI-78, gamma-secretase subunit APH-1A, Presenilin-stabilization factor, PSF | |
| APH1A | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, monkey, mouse, cow and dog, 94% homologous in rat and 88% homologous in chicken. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuroscience, Stem Cell Markers | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 51107 | |
| A portion of amino acids 80-145 of human APH1A was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title