missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APH1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27176
This item is not returnable.
View return policy
Description
APH1A Polyclonal specifically detects APH1A in Human, Bovine, Canine, Equine, Opossum, Primate samples. It is validated for Western Blot.
Specifications
| APH1A | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| APH1A | |
| A portion of amino acids 80-145 of human APH1A was used as the immunogen. | |
| 0.1 mg | |
| GPCR, Neuroscience, Stem Cell Markers | |
| 51107 | |
| Human, Bovine, Canine, Equine, Opossum, Primate | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 4-6ug/ml | |
| 6530402N02Rik, anterior pharynx defective 1 homolog A (C. elegans), APH-1, APH-1A, aph-1alpha, CGI-78, gamma-secretase subunit APH-1A, Presenilin-stabilization factor, PSF | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, monkey, mouse, cow and dog, 94% homologous in rat and 88% homologous in chicken. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction