missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APEX™ Antibody Labeling Kits

Product Code. 10565664
Change view
Click to view available options
Quantity:
5 Reactions
Excitation/Emission:
410/455
490/525
555/580
578/603
590/617
650/665
Label or Dye:
Alexa Fluor 488
Alexa Fluor 555
Alexa Fluor 568
Alexa Fluor 594
Alexa Fluor 647
Biotin-XX
Pacific Blue
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Excitation/Emission Label or Dye unitSize
10565664 5 Reactions 650/665 Alexa Fluor 647 Each
10676255 5 Reactions 490/525 Alexa Fluor 488 Each
10287124 5 Reactions 555/580 Alexa Fluor 555 Each
10482555 5 Reactions 590/617 Alexa Fluor 594 Each
10469864 5 Reactions 410/455 Pacific Blue Each
10440430 5 Reactions 578/603 Alexa Fluor 568 Each
10477010 5 Reactions - Biotin-XX Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10565664 Supplier Invitrogen™ Supplier No. A10475

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Generate ready-to-use, Alexa Fluor-labeled antibodies even in the presence of contaminants with APEX Alexa Fluor Antibody Labeling kits.

Invitrogen APEX Antibody Labeling kits enable you to covalently label small quantities of IgG antibody (10–20 μg) with Alexa Fluor dyes or biotin, even in the presence of contaminants, stabilizing proteins, or amine-containing buffer (e.g., BSA, Tris, or glycine). APEX Antibody Labeling kits feature a resin tip that captures the IgG antibody, enabling the removal of contaminants prior to the labeling reaction with the conjugate of choice. After the application of the amine-reactive fluorophore to the IgG antibody bound on the resin, a fluorescent IgG conjugate is formed and subsequently eluted from the resin using elution buffer. Each kit contains everything needed for five labeling reactions. Working IgG antibody conjugate can be achieved in about 2.5 hours (with approximately 15 minutes of hands-on time).

Invitrogen APEX Antibody Labeling kits enable the covalent attachment of a fluorophore or biotin to small amounts of IgG antibody (∼10–20 μg), even in contaminated mixtures or solutions of biological samples. They are ideal for the efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions. Labeled antibodies are ready for use in imaging or flow cytometry applications in as little as 2.5 hours with little hands-on time.

Important features of APEX Antibody Labeling kits:
· Labeled antibodies typically ready to use in 2.5 hours (∼15 minutes hands-on time)
· Designed to label 10–20 μg of IgG
· Covalent attachment of Alexa Fluor dyes or biotin
· Compatible with contaminating proteins or stabilizers like BSA
· Choose from Alexa Fluor 488, 555, 568, 594, and 647 dyes, Pacific Blue dye, or Biotin-XX

Contaminating proteins or protein stabilizers are not a problem
Many IgG antibodies are often available only in small quantities and packaged with stabilizing proteins, such as BSA, or other contaminants that can interfere with the amine-reactive labeling reagents. The APEX Antibody Labeling kits avoid this issue by utilizing a solid-phase labeling technique that captures the IgG antibody on a specialized resin inside the APEX antibody labeling tip. Contaminants are simply eluted through the tip prior to the application of the amine-reactive label.

Better results and workflows with primary labeled antibodies
A primary antibody directly labeled with a fluorophore often produces lower background fluorescence and less nonspecific binding. Further, multiple primary antibodies of the same isotype or derived from the same species can easily be used in the same experiment if they are directly labeled with compatible fluorophores.

Customization is possible
Can’t find what you’re looking for in our stocked list? We can prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 9001:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

Kit Contents Contains Reactive Fluorescent Label, 5 vials; APEX antibody labeling tips, 5 Ea.; Wash Buffer, 1.8 mL; Dimethylsulfoxide (DMSO), 100 μL; Labeling Buffer, 100 μL; Neutralization Buffer, 100 μL; Elution Buffer, 1 mL; Elution Syringe, 1 Ea.
Quantity 5 Reactions
Content And Storage Store at 2°C to 6°C, do not freeze, desiccate and protect from light.
Excitation/Emission 650/665
Labeling Scale 10 to 20 μg
Product Line APEX
Chemical Reactivity Amine
Label or Dye Alexa Fluor 647
Color Far-Red
Labeling Target IgG
Label Type Alexa Fluor Dyes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.