missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ APET Liners for Safety Trays

Product Code. 12347207 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
260 x 460mm
340 x 540mm
460 x 700mm
540 x 570mm
540 x 680mm
Unit Size:
Pack of 25

Product Code. 12347207

Only null left
Add to basket
Add to basket

Environmental friendly, easy to clean and maintain for routine work and storage in safe areas.

  • Clears up spillages easily
  • Environmental friendly

Specifications

Dimensions (L x W) 460 x 700mm
Length (Metric) 460 mm
Material APET
Type Liners
Width (Metric) 700 mm
For Use With Radiation, Biohazard, General purpose Trays
No. per Pack 25
Shape Rectangular
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.