missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APCDD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£187.00 - £410.00
Specifications
| Antigen | APCDD1 |
|---|---|
| Concentration | 1.3 mg/mL |
| Dilution | Western Blot 0.5 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18179189
|
Novus Biologicals
NB110-92756SS |
0.025 mL |
£187.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18784983
|
Novus Biologicals
NB110-92756 |
0.1 mL |
£410.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APCDD1 Polyclonal specifically detects APCDD1 in Human, Mouse, Chicken, Reptile samples. It is validated for Western Blot.Specifications
| APCDD1 | |
| Western Blot 0.5 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Cancer | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| adenomatosis polyposis coli down-regulated 1, Adenomatosis polyposis coli down-regulated 1 protein, B7323, DRAPC1, protein APCDD1 | |
| APCDD1 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, chicken and turtle. |
| 1.3 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Chicken, Reptile | |
| Q8J025 | |
| 147495 | |
| Synthetic peptide made to an internal portion of human APCDD1 (within residues 400-500). [Swiss-Prot# Q8J025] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 58 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title