missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APCDD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB110-92756
This item is not returnable.
View return policy
Description
APCDD1 Polyclonal specifically detects APCDD1 in Human, Mouse, Chicken, Reptile samples. It is validated for Western Blot.
Specifications
| APCDD1 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| adenomatosis polyposis coli down-regulated 1, Adenomatosis polyposis coli down-regulated 1 protein, B7323, DRAPC1, protein APCDD1 | |
| Rabbit | |
| 58 kDa | |
| 0.1 mL | |
| Cancer | |
| 147495 | |
| Human, Mouse, Chicken, Reptile | |
| IgG |
| Western Blot | |
| 1.3 mg/mL | |
| Western Blot 0.5 ug/ml | |
| Q8J025 | |
| APCDD1 | |
| Synthetic peptide made to an internal portion of human APCDD1 (within residues 400-500). [Swiss-Prot# Q8J025] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, chicken and turtle. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction