missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apc1 Antibody (7G9C7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | Apc1 |
|---|---|
| Clone | 7G9C7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18200323
|
Novus Biologicals
NBP2-61887 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647468
|
Novus Biologicals
NBP2-61887-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Apc1 Monoclonal specifically detects Apc1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| Apc1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| anaphase promoting complex subunit 1, anaphase-promoting complex 1 (meiotic checkpoint regulator), anaphase-promoting complex subunit 1, APC1Testis-specific gene 24 protein, Cyclosome subunit 1, Mitotic checkpoint regulator, TSG24MCPR | |
| ANAPC1 | |
| IgG1 | |
| Protein G purified |
| 7G9C7 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Cell Biology, Cell Cycle and Replication, Phospho Specific, Ubiquitin Proteasome Pathway | |
| 64682 | |
| Purified recombinant fragment of human ANAPC1 (AA: 12-155) expressed in E. Coli. | |
| Primary | |
| 216 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title