missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apc1 Antibody (7G9C7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61887
This item is not returnable.
View return policy
Description
Apc1 Monoclonal specifically detects Apc1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| Apc1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| anaphase promoting complex subunit 1, anaphase-promoting complex 1 (meiotic checkpoint regulator), anaphase-promoting complex subunit 1, APC1Testis-specific gene 24 protein, Cyclosome subunit 1, Mitotic checkpoint regulator, TSG24MCPR | |
| Mouse | |
| 216 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 7G9C7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ANAPC1 | |
| Purified recombinant fragment of human ANAPC1 (AA: 12-155) expressed in E. Coli. | |
| Protein G purified | |
| Cell Biology, Cell Cycle and Replication, Phospho Specific, Ubiquitin Proteasome Pathway | |
| 64682 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction