missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC-PCI Complex Mouse anti-Human, Clone: M36, Novus Biologicals™
Mouse Monoclonal Antibody
£175.00 - £427.00
Specifications
| Antigen | APC-PCI Complex |
|---|---|
| Clone | M36 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18107918
|
Novus Biologicals
NBP2-41380-0.025MG |
0.025 mg |
£175.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642038
|
Novus Biologicals
NBP2-41380 |
100 μg |
£427.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APC-PCI Complex Monoclonal specifically detects APC-PCI Complex in Human samples. It is validated for ELISA.Specifications
| APC-PCI Complex | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Protein A or G purified |
| M36 | |
| ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mixture of PCI in complex with PSA (approx. 80%) and PCI cleaved from such a complex (approx. 20%). | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title