missing translation for 'onlineSavingsMsg'
Learn More

APC-PCI Complex Mouse anti-Human, Clone: M36, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-41380-0.025MG

 View more versions of this product

Product Code. 18107918

  • £175.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

APC-PCI Complex Monoclonal specifically detects APC-PCI Complex in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

APC-PCI Complex
Monoclonal
1 mg/mL
ELISA 1:100-1:2000
Mouse
Protein A or G purified
RUO
Antibody is specific for APC:PCI complex and cleaved PCI but has little reactivity towards native PCI. The dissociation constants (as determined by standard surface plasmon resonance technique) are: Kd= 4x10e-10 M for PCI in complex with APC and 2x10e-10 M for cleaved PCI. The value of Kd for native PCI is too low to measure, i.e. > 10e-5 M.
Store at 4C in the dark.
IgG1 κ
ELISA
M36
Unconjugated
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
Mixture of PCI in complex with PSA (approx. 80%) and PCI cleaved from such a complex (approx. 20%).
0.025 mg
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only