missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC Antibody (Ali 12-28) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
£169.00
Specifications
| Antigen | APC |
|---|---|
| Clone | ALi 12-28 |
| Concentration | 1.2 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
Description
APC Monoclonal specifically detects APC in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| APC | |
| 1.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P25054 | |
| 324 | |
| N-terminal APC-MBP fusion protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| ALi 12-28 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Metastasis, Tumor Suppressors, Wnt Signaling Pathway | |
| PBS with 0.02% Sodium Azide | |
| adenomatosis polyposis coli, adenomatosis polyposis coli tumor suppressor, adenomatous polyposis coli, adenomatous polyposis coli protein, BTPS2, Deleted in polyposis 2.5, DP2.5DP2, DP3, FPC, GS, Protein APC | |
| APC | |
| IgG1 | |
| Protein A purified | |
| Ali 12-28 detects both normal and mutant-truncated forms of the APC. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title