missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC Antibody (Ali 12-28) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-667-0.025mg
This item is not returnable.
View return policy
Description
APC Monoclonal specifically detects APC in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| APC | |
| Monoclonal | |
| 1.2 mg/mL | |
| PBS with 0.02% Sodium Azide | |
| adenomatosis polyposis coli, adenomatosis polyposis coli tumor suppressor, adenomatous polyposis coli, adenomatous polyposis coli protein, BTPS2, Deleted in polyposis 2.5, DP2.5DP2, DP3, FPC, GS, Protein APC | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Ali 12-28 detects both normal and mutant-truncated forms of the APC. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| ELISA | |
| ALi 12-28 | |
| Unconjugated | |
| P25054 | |
| APC | |
| N-terminal APC-MBP fusion protein | |
| 0.025 mg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Metastasis, Tumor Suppressors, Wnt Signaling Pathway | |
| 324 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction