missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APAF1 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5145497

 View more versions of this product

Product Code. 17900073

  • £341.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

APAF1 gene encodes a cytoplasmic protein that initiates apoptosis. This protein contains several copies of the WD-40 domain, a caspase recruitment domain (CARD), and an ATPase domain (NB-ARC). Upon binding cytochrome c and dATP, this protein forms an oligomeric apoptosome. The apoptosome binds and cleaves caspase 9 preproprotein, releasing its mature, activated form. Activated caspase 9 stimulates the subsequent caspase cascade that commits the cell to apoptosis. Alternative splicing results in several transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

APAF1
Polyclonal
Unconjugated
APAF1
6230400I06Rik; APAF1; APAF-1; Apaf1l; apoptotic peptidase activating factor 1; apoptotic protease activating factor 1; apoptotic protease-activating factor 1; CED4; DKFZp781B1145; fog; forebrain overgrowth; KIAA0413; mKIAA0413
Rabbit
Antigen affinity chromatography, Protein A
RUO
11783, 78963
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
O88879, Q9EPV5
APAF1
Recombinant Mouse APAF1 protein, Arg872-Phe1135 (Accession #O88879).
200 μL
Primary
Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.