missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APAF-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£134.00
Specifications
| Antigen | APAF-1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 1 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 1 μg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
APAF-1 Polyclonal specifically detects APAF-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| APAF-1 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 1 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 1 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Tumor Suppressors | |
| PBS with 0.02% Sodium Azide | |
| APAF-1apoptotic protease activating factor 1, apoptotic peptidase activating factor, apoptotic peptidase activating factor 1, apoptotic protease activating factor, apoptotic protease-activating factor 1, CED4, DKFZp781B1145, KIAA0413 | |
| APAF1 | |
| IgG | |
| Affinity Purified | |
| 130 kDa |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| AAC51678 | |
| 317 | |
| Antibody was raised against a peptide corresponding to amino acids near the amino terminus of human Apaf1. The sequences of the immunogenic peptide are identical between human and mouse. The immunogen is located within the first 50 amino acids of Apaf1. Amino Acid Squence: HREALEKDIKTSYIMDH | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title