missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APAF-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-77000-0.025mg
This item is not returnable.
View return policy
Description
APAF-1 Polyclonal specifically detects APAF-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| APAF-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| APAF-1apoptotic protease activating factor 1, apoptotic peptidase activating factor, apoptotic peptidase activating factor 1, apoptotic protease activating factor, apoptotic protease-activating factor 1, CED4, DKFZp781B1145, KIAA0413 | |
| Rabbit | |
| 130 kDa | |
| 0.025 mg | |
| Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Tumor Suppressors | |
| 317 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 1 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 1 μg/mL | |
| AAC51678 | |
| APAF1 | |
| Antibody was raised against a peptide corresponding to amino acids near the amino terminus of human Apaf1. The sequences of the immunogenic peptide are identical between human and mouse. The immunogen is located within the first 50 amino acids of Apaf1. Amino Acid Squence: HREALEKDIKTSYIMDH | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction