missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AP3M1 Recombinant Rabbit Monoclonal Antibody (24GB6160)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557337
This item is not returnable.
View return policy
Description
AP3M1 Recombinant Monoclonal Antibody for Western Blot
The AP3 complex is a heterotetramer composed of two large adaptins (AP3D1, AP3B1 or AP3B2), a medium adaptin (AP3M1 or AP3M2) and a small adaptin (APS1 or AP3S2). The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes. AP3M1 is highly homologous to AP3M2 but is the preferred binding partner to the YQRL motif from the cytosolic domain of TGN38. AP3M1 has also been shown to interact with the HIV-1 virulence protein Nef.
Specifications
| AP3M1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Ap3m1 | |
| 1200013D09Rik; adapter-related protein complex 3 mu-1 subunit; adapter-related protein complex 3 subunit mu-1; adaptor protein complex 3 Mu3A; adaptor related protein complex 3 mu 1 subunit; adaptor related protein complex 3 subunit mu 1; adaptor-related protein complex 3 subunit mu-1; adaptor-related protein complex 3, mu 1 subunit; adaptor-related protein complex AP-3 mu 1 subunit; adaptor-related protein complex AP-3, mu 1 subunit; ap3; AP-3 adapter complex mu3A subunit; AP-3 adaptor complex mu3A subunit; AP-3 complex subunit mu-1; AP-3 complex subunit mu-1-like protein; Ap3m1; C78982; clathrin adaptor complex AP3, mu-3A subunit; clathrin adaptor protein mu3A; clathrin assembly protein assembly protein complex 1 medium chain homolog 1; clathrin assembly protein assembly protein complex 3 mu-1 medium chain; clathrin coat assembly protein AP47 homolog 1; clathrin coat-associated protein AP47 homolog 1; clathrin-associated adaptor protein homolog (p47A); golgi adaptor AP-1 47 kDa protein homolog 1; HA1 47 kDa subunit homolog 1; hypothetical protein; Mu3A-adaptin; mu-adaptin 3A; P47a; R75378; RCJMB04_1h22; Unknown (protein for MGC:137644) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 171126, 26985, 55946 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 24GB6160 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P53676, Q9JKC8, Q9Y2T2 | |
| Ap3m1 | |
| A synthesized peptide derived from human AP3M1 (aa 146-159). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction