missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AP3M1 Recombinant Rabbit Monoclonal Antibody (24GB6160)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen AP3M1
Clone 24GB6160
Content And Storage -20°C
Applications Western Blot
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275872
GREENER_CHOICE
View Documents
Invitrogen™
MA557337
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275475
View Documents
Invitrogen™
MA557338
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AP3M1 Recombinant Monoclonal Antibody for Western Blot

The AP3 complex is a heterotetramer composed of two large adaptins (AP3D1, AP3B1 or AP3B2), a medium adaptin (AP3M1 or AP3M2) and a small adaptin (APS1 or AP3S2). The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes. AP3M1 is highly homologous to AP3M2 but is the preferred binding partner to the YQRL motif from the cytosolic domain of TGN38. AP3M1 has also been shown to interact with the HIV-1 virulence protein Nef.
Specifications

Specifications

AP3M1
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
Ap3m1
1200013D09Rik; adapter-related protein complex 3 mu-1 subunit; adapter-related protein complex 3 subunit mu-1; adaptor protein complex 3 Mu3A; adaptor related protein complex 3 mu 1 subunit; adaptor related protein complex 3 subunit mu 1; adaptor-related protein complex 3 subunit mu-1; adaptor-related protein complex 3, mu 1 subunit; adaptor-related protein complex AP-3 mu 1 subunit; adaptor-related protein complex AP-3, mu 1 subunit; ap3; AP-3 adapter complex mu3A subunit; AP-3 adaptor complex mu3A subunit; AP-3 complex subunit mu-1; AP-3 complex subunit mu-1-like protein; Ap3m1; C78982; clathrin adaptor complex AP3, mu-3A subunit; clathrin adaptor protein mu3A; clathrin assembly protein assembly protein complex 1 medium chain homolog 1; clathrin assembly protein assembly protein complex 3 mu-1 medium chain; clathrin coat assembly protein AP47 homolog 1; clathrin coat-associated protein AP47 homolog 1; clathrin-associated adaptor protein homolog (p47A); golgi adaptor AP-1 47 kDa protein homolog 1; HA1 47 kDa subunit homolog 1; hypothetical protein; Mu3A-adaptin; mu-adaptin 3A; P47a; R75378; RCJMB04_1h22; Unknown (protein for MGC:137644)
Ap3m1
Primary
Affinity chromatography
24GB6160
Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P53676, Q9JKC8, Q9Y2T2
171126, 26985, 55946
A synthesized peptide derived from human AP3M1 (aa 146-159).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.