missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP3D1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £302.00
Specifications
| Antigen | AP3D1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-5ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18123743
|
Novus Biologicals
NBP2-27158SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18111114
|
Novus Biologicals
NBP2-27158 |
0.1 mg |
£302.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AP3D1 Polyclonal specifically detects AP3D1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AP3D1 | |
| Western Blot 2-5ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 8943 | |
| A portion of amino acids 530-580 of human AP3 Delta was used as the immunogen for the antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Adapter-related protein complex 3 subunit delta-1, adaptin, delta, adaptor-related protein complex 3, delta 1 subunit, ADTD, AP-3 complex delta subunit, partial CDS, AP-3 complex subunit delta, AP-3 complex subunit delta-1, delta adaptin, Delta-adaptin, delta-adaptin, partial CDS, hBLVR, subunit of putative vesicle coat adaptor complex AP-3 | |
| AP3D1 | |
| IgG | |
| Protein A purified | |
| The AP3 amino acid sequence used as immunogen for this antibody is 100% homologous in human, chimpanzee, orangutan, new world monkey, rhesus monkey, and 63% homologous in panda, Carolina anole, opossum, and 58% homologous in taipan, zebra finch, horse, dog, pig and 53% homologous in cow, rat, mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title