missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP3D1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27158
This item is not returnable.
View return policy
Description
AP3D1 Polyclonal specifically detects AP3D1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AP3D1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| AP3D1 | |
| A portion of amino acids 530-580 of human AP3 Delta was used as the immunogen for the antibody. | |
| 0.1 mg | |
| Primary | |
| The AP3 amino acid sequence used as immunogen for this antibody is 100% homologous in human, chimpanzee, orangutan, new world monkey, rhesus monkey, and 63% homologous in panda, Carolina anole, opossum, and 58% homologous in taipan, zebra finch, horse, dog, pig and 53% homologous in cow, rat, mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 2-5ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml | |
| Adapter-related protein complex 3 subunit delta-1, adaptin, delta, adaptor-related protein complex 3, delta 1 subunit, ADTD, AP-3 complex delta subunit, partial CDS, AP-3 complex subunit delta, AP-3 complex subunit delta-1, delta adaptin, Delta-adaptin, delta-adaptin, partial CDS, hBLVR, subunit of putative vesicle coat adaptor complex AP-3 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 8943 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction