missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP3B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00
Specifications
| Antigen | AP3B2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18514334
|
Novus Biologicals
NB100-74370-0.025mg |
0.025 mg | |||||||
Description
AP3B2 Polyclonal specifically detects AP3B2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| AP3B2 | |
| Western Blot 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q13367, Q9JME5 | |
| 8120 | |
| Syntehtic peptides corresponding to residues L(260) L E E N (A/P) E K A F Y G S E E(274) of human and mouse beta NAP. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 1.0 mg/ml BSA with 0.05% Sodium Azide | |
| Adapter-related protein complex 3 subunit beta-2, Adaptor protein complex AP-3 subunit beta-2, adaptor-related protein complex 3, beta 2 subunit, beta-3B-adaptin, Clathrin assembly protein complex 3 beta-2 large chain, DKFZp686D17136, NAPTBAP-3 complex subunit beta-2, Neuronal adaptin-like protein, beta-subunit, Neuron-specific vesicle coat protein beta-NAP | |
| AP3B2 | |
| IgG | |
| Immunogen affinity purified | |
| beta NAP |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title