missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP3B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74370-0.025mg
This item is not returnable.
View return policy
Description
AP3B2 Polyclonal specifically detects AP3B2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| AP3B2 | |
| Polyclonal | |
| Unconjugated | |
| 1.0 mg/ml BSA with 0.05% Sodium Azide | |
| Adapter-related protein complex 3 subunit beta-2, Adaptor protein complex AP-3 subunit beta-2, adaptor-related protein complex 3, beta 2 subunit, beta-3B-adaptin, Clathrin assembly protein complex 3 beta-2 large chain, DKFZp686D17136, NAPTBAP-3 complex subunit beta-2, Neuronal adaptin-like protein, beta-subunit, Neuron-specific vesicle coat protein beta-NAP | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8120 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500 | |
| Q13367, Q9JME5 | |
| AP3B2 | |
| Syntehtic peptides corresponding to residues L(260) L E E N (A/P) E K A F Y G S E E(274) of human and mouse beta NAP. | |
| 0.025 mg | |
| Primary | |
| beta NAP | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction