missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP1M2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | AP1M2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
AP1M2 Polyclonal specifically detects AP1M2 in Human samples. It is validated for Western Blot.Specifications
| AP1M2 | |
| Polyclonal | |
| Rabbit | |
| Q9Y6Q5 | |
| 10053 | |
| Synthetic peptides corresponding to AP1M2 (adaptor-related protein complex 1, mu 2 subunit) The peptide sequence was selected from the N terminal of AP1M2 (NP_005489). Peptide sequence: MSASAVFILDVKGKPLISRNYKGDVAMSKIEHFMPLLVQREEEGALAPLL. | |
| Primary | |
| 48 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Adaptor protein complex AP-1 mu-2 subunit, Adaptor-related protein complex 1 mu-2 subunit, adaptor-related protein complex 1, mu 2 subunit, AP-1 complex subunit mu-2, AP1-mu2, AP-mu chain family member mu1B, Clathrin assembly protein complex 1 medium chain 2, clathrin coat assembly protein AP47 2, clathrin coat associated protein AP47 2, clathrin-associated adaptor medium chain mu2, golgi adaptor AP-1 47 kDa protein, Golgi adaptor HA1/AP1 adaptin mu-2 subunit, HA1 47 kDa subunit 2, HSMU1B, MU1B, MU-1B, mu1B-adaptin, mu2, Mu-adaptin 2 | |
| AP1M2 | |
| IgG | |
| This product is specific to Subunit or Isoform: mu-2. |
Spot an opportunity for improvement?Share a Content Correction
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts