missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP1M2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57035
This item is not returnable.
View return policy
Description
AP1M2 Polyclonal specifically detects AP1M2 in Human samples. It is validated for Western Blot.
Specifications
| AP1M2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Adaptor protein complex AP-1 mu-2 subunit, Adaptor-related protein complex 1 mu-2 subunit, adaptor-related protein complex 1, mu 2 subunit, AP-1 complex subunit mu-2, AP1-mu2, AP-mu chain family member mu1B, Clathrin assembly protein complex 1 medium chain 2, clathrin coat assembly protein AP47 2, clathrin coat associated protein AP47 2, clathrin-associated adaptor medium chain mu2, golgi adaptor AP-1 47 kDa protein, Golgi adaptor HA1/AP1 adaptin mu-2 subunit, HA1 47 kDa subunit 2, HSMU1B, MU1B, MU-1B, mu1B-adaptin, mu2, Mu-adaptin 2 | |
| Rabbit | |
| 48 kDa | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: mu-2. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9Y6Q5 | |
| AP1M2 | |
| Synthetic peptides corresponding to AP1M2 (adaptor-related protein complex 1, mu 2 subunit) The peptide sequence was selected from the N terminal of AP1M2 (NP_005489). Peptide sequence: MSASAVFILDVKGKPLISRNYKGDVAMSKIEHFMPLLVQREEEGALAPLL. | |
| Affinity purified | |
| RUO | |
| 10053 | |
| Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction