Learn More
Novus Biologicals™ Antibody Labeling Confirmation Control Kit
Product Code. 18038334 Shop All Bio Techne Products
Click to view available options
Quantity:
30 Tests
Unit Size:
30 tests
Product Code. 18038334
Brand: Novus Biologicals™ 40000030
Allows to confirm successful conjugation of antibody.
  • Chromatography test
  • Compatible with conjugation technology that uses colored labels
  • Requires small volumes (40μl of diluted conjugate)
TRUSTED_SUSTAINABILITY
Specifications
Quantity 30 Tests
Product Type Antibody detection confirmation kit