Learn More
Invitrogen™ Antibody Diluent
Antibody Diluent
Brand: Invitrogen™ 003218
Additional Details : Weight : 0.63500kg
Description
Ready-to-use diluent with BSA (for stabilizing the antibody) and preservative.“This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”
Specifications
Ready-to-Use Antibody Diluent | |
Antibody Diluent, 500mL, Liquid, Ready-to-Use Diluent, 26628-22-8 CAS Number, 0.1% Sodium Azide Preservative, BSA Stabilizer Additives, Highly Purified Grade, Wet Ice Shipping Condition, 2 to 8°C Storage Temperature, For In-vitro Diagnostic Use | |
2°C to 8°C | |
Liquid |
Cell analysis, cellular imaging, immunohistochemistry (IHC), IHC staining and detection | |
For in-vitro diagnostic use | |
500 mL |
For In Vitro Diagnostic Use.