missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antibody Diluent
Antibody Diluent
£89.75 - £123.00
Specifications
Product Type | Ready-to-Use Antibody Diluent |
---|---|
For Use With (Application) | Cell analysis, cellular imaging, immunohistochemistry (IHC), IHC staining and detection |
Regulatory Status | For in-vitro diagnostic use |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10414753
|
Invitrogen™
003118 |
250 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15908982
|
Invitrogen™
003218 |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Ready-to-use diluent with BSA (for stabilizing the antibody) and preservative.“This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”
Specifications
Ready-to-Use Antibody Diluent | |
For in-vitro diagnostic use | |
Liquid |
Cell analysis, cellular imaging, immunohistochemistry (IHC), IHC staining and detection | |
2°C to 8°C |
For In Vitro Diagnostic Use.