missing translation for 'onlineSavingsMsg'
Learn More

XIAP Mouse, Unlabeled, Clone: 48, BD

Mouse Monoclonal Antibody

£163.84 - £298.29

Specifications

Antigen XIAP
Clone 48
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15865629
View Documents
BD Biosciences
610762
50 μg
ONLINE EXCLUSIVE PRICE
£171.00
£163.84 / 1 set
Save £7.16
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15875629
View Documents
BD Biosciences
610763
150 μg
ONLINE EXCLUSIVE PRICE
£311.00
£298.29 / 1 set
Save £12.71
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Apoptosis is a genetically programmed, selective process of cell death that occurs during normal cell differentiation and development of multicellular organisms. Viruses depend on the biosynthetic machinery of their host cell for the production of progeny and survival. Therefore, many viruses encode proteins that protect the cell from apoptosis. hILP ( h uman I AP- l ike p rotein) is a human homologue of the viral IAP ( I nhibitor of A poptosis P rotein). hILP is a widely expressed cytoplasmic protein of 497 amino acids with three BIR (Baculovirus IAP repeats) domains and a C-terminal RING finger domain. hILP-transfected cells are protected against the apoptotic effects of Sindbis virus infection and ICE (interleukin-1 β; converting enzyme) expression. This product is sold under license from Aegera Therapeutics, Inc.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications

Specifications

XIAP
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
hILP
48
Western Blot
Unconjugated
RUO
Frog, Human, Mouse, Rat
Human hILP aa. 268-426
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.