missing translation for 'onlineSavingsMsg'
Learn More

XIAP Mouse, Unlabeled, Clone: 48, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610762

 View more versions of this product

Product Code. 15865629

  • ONLINE EXCLUSIVE PRICE
    £171.00
    £163.84 / 1 set
    Save £7.16
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Apoptosis is a genetically programmed, selective process of cell death that occurs during normal cell differentiation and development of multicellular organisms. Viruses depend on the biosynthetic machinery of their host cell for the production of progeny and survival. Therefore, many viruses encode proteins that protect the cell from apoptosis. hILP ( h uman I AP- l ike p rotein) is a human homologue of the viral IAP ( I nhibitor of A poptosis P rotein). hILP is a widely expressed cytoplasmic protein of 497 amino acids with three BIR (Baculovirus IAP repeats) domains and a C-terminal RING finger domain. hILP-transfected cells are protected against the apoptotic effects of Sindbis virus infection and ICE (interleukin-1 β; converting enzyme) expression. This product is sold under license from Aegera Therapeutics, Inc.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

XIAP
Monoclonal
250μg/mL
hILP
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
48
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human hILP aa. 268-426
50 μg
Cell Biology
Frog, Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.