Learn More
TrkB Mouse anti-Rat, Unlabeled, Clone: 47, BD
Mouse Monoclonal Antibody
£152.16 - £190.20
Specifications
| Antigen | TrkB |
|---|---|
| Clone | 47 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15855259
|
BD Biosciences
610101 |
50 μg |
£159.00 £152.16 / 1 set Save £6.84 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15865259
|
BD Biosciences
610102 |
150 μg |
£198.00 £190.20 / 1 set Save £7.80 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The full-length TrkB gene has been reported to encode for a 145kDa glycosylated transmembrane tyrosine kinase and neurotrophin receptor. The same gene also has been reported to encode for a 95kDa glycoprotein that is identical to gp145 (TrkB) at the extracellular domain and transmembrane portion but lacks the intracellular portion. TrkB has been observable in a range of 116-145kDa and 70-95kDa due to various TrkB maturation states, subcellular localizations, and glycosylation states. TrkB belongs to a family of tyrosine kinases that include the TrkA proto-oncogene and TrkC. All have an extracellular ligand-binding domain, a transmembrane region, and intracellular kinase and autophosphorylation domains. TrkB binds the neurotrophins NT3 and NT4/5, as well as brain-derived neurotrophic factor (BDNF), a peptide that helps motor neuron survival and repair. The TrkB tyrosine kinase is activated upon binding to BDNF resulting in autophosphorylation of residues Y670, Y674 and Y675 and the subsequent association of several intracellular proteins like PLCγ, Shc, and PI3-Kinase. TrkB is widely expressed in cells of neuroepithelium and neural crest origin. Some of these include motor neurons, dopamine-producing neurons, and neurons which release γ-aminobutyric acid in the substantia nigra, neocortex, and hippocampus. The two TrkB gene products are differentially expressed in regions of the adult brain.
Host Species: Mouse
Clone: 47
Isotype: IgG1
Species Reactivity: Rat, Human
Immunogen: Human TrkB aa. 156-322
Formula Weight [Chemical]: 116-145 & 70-95kDa
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| TrkB | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified | |
| Neurotrophic Tyrosine Kinase Receptor Type 2; NTRK2 |
| 47 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Rat | |
| Human TrkB aa. 156-322 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.