missing translation for 'onlineSavingsMsg'
Learn More

TrkB Mouse anti-Rat, Unlabeled, Clone: 47, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610102

 View more versions of this product

Product Code. 15865259

  • ONLINE EXCLUSIVE PRICE
    £198.00
    £190.20 / 1 set
    Save £7.80
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The full-length TrkB gene has been reported to encode for a 145kDa glycosylated transmembrane tyrosine kinase and neurotrophin receptor. The same gene also has been reported to encode for a 95kDa glycoprotein that is identical to gp145 (TrkB) at the extracellular domain and transmembrane portion but lacks the intracellular portion. TrkB has been observable in a range of 116-145kDa and 70-95kDa due to various TrkB maturation states, subcellular localizations, and glycosylation states. TrkB belongs to a family of tyrosine kinases that include the TrkA proto-oncogene and TrkC. All have an extracellular ligand-binding domain, a transmembrane region, and intracellular kinase and autophosphorylation domains. TrkB binds the neurotrophins NT3 and NT4/5, as well as brain-derived neurotrophic factor (BDNF), a peptide that helps motor neuron survival and repair. The TrkB tyrosine kinase is activated upon binding to BDNF resulting in autophosphorylation of residues Y670, Y674 and Y675 and the subsequent association of several intracellular proteins like PLCγ, Shc, and PI3-Kinase. TrkB is widely expressed in cells of neuroepithelium and neural crest origin. Some of these include motor neurons, dopamine-producing neurons, and neurons which release γ-aminobutyric acid in the substantia nigra, neocortex, and hippocampus. The two TrkB gene products are differentially expressed in regions of the adult brain.

Host Species: Mouse
Clone: 47
Isotype: IgG1
Species Reactivity: Rat, Human
Immunogen: Human TrkB aa. 156-322
Formula Weight [Chemical]: 116-145 & 70-95kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TrkB
Monoclonal
250μg/mL
Neurotrophic Tyrosine Kinase Receptor Type 2; NTRK2
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
47
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human TrkB aa. 156-322
150 μg
Cell Biology
Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.