missing translation for 'onlineSavingsMsg'
Learn More

TetR Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£175.00 - £381.00

Specifications

Applications Western Blot
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18797873
View Documents
Novus Biologicals
NBP1-78424SS
£175.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18210256
View Documents
Novus Biologicals
NBP1-78424
0.1 mL
£381.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TetR Polyclonal specifically detects TetR in Non-species specific samples. It is validated for Western Blot.
Specifications

Specifications

Western Blot
Rabbit
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.