missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TetR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78424
This item is not returnable.
View return policy
Description
TetR Polyclonal specifically detects TetR in Non-species specific samples. It is validated for Western Blot.
Specifications
| TetR | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| pSC101 tetR, Tet Repressor, Tet Repressor Protein, TetR, tetracycline repressor protein | |
| A synthetic peptide made to an N-terminal portion of Escherichia coli TetR protein (between residues 1-50) [UniProt P04483] | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 4924774 | |
| Non-species specific | |
| IgG |
| Western Blot | |
| 1.13 mg/mL | |
| Western Blot 1:1000 | |
| P04483 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is used to monitor the regulation of gene expression in bacteria cells. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction