Learn More
Rb2 Mouse, Unlabeled, Clone: 10, BD
Mouse Monoclonal Antibody
£219.86 - £396.51
Specifications
| Antigen | Rb2 |
|---|---|
| Clone | 10 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15835369
|
BD Biosciences
610261 |
50 μg |
£229.00 £219.86 / 1 set Save £9.14 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15845369
|
BD Biosciences
610262 |
150 μg |
£412.00 £396.51 / 1 set Save £15.49 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The pRb2 protein shows a high degree of identity with pRb and the related p107 protein. Both pRb and p107 bind to the adenovirus E1A, SV40 large T antigen, and papillomavirus E7 viral proteins. This binding initiates the release of transcription factors which are required for the expression of cell cycle-regulated genes. pRb, pRb2, and p107 interact with a conserved motif in these three viral proteins. In the E1A protein, this area is known as transforming domain 2, which is required for growth activation. The E1A-binding domain in pRb and p107 is a conserved motif known as the “pocket region” which consists of conserved A and B regions separated by non-conserved spacers of different sizes in pRb and p107. It is the pocket regions of pRb and p107 that associate with the E2F transcription factor. pRb2 also contains the conserved pocket region suggesting that it has functional similarities to pRb and p107.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| Rb2 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG2a | |
| Affinity Purified |
| 10 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Chicken, Drosophila, Human, Mouse, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.