missing translation for 'onlineSavingsMsg'
Learn More

Rb2 Mouse, Unlabeled, Clone: 10, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610261

 View more versions of this product

Product Code. 15835369

  • ONLINE EXCLUSIVE PRICE
    £229.00
    £219.86 / 1 set
    Save £9.14
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The pRb2 protein shows a high degree of identity with pRb and the related p107 protein. Both pRb and p107 bind to the adenovirus E1A, SV40 large T antigen, and papillomavirus E7 viral proteins. This binding initiates the release of transcription factors which are required for the expression of cell cycle-regulated genes. pRb, pRb2, and p107 interact with a conserved motif in these three viral proteins. In the E1A protein, this area is known as transforming domain 2, which is required for growth activation. The E1A-binding domain in pRb and p107 is a conserved motif known as the “pocket region” which consists of conserved A and B regions separated by non-conserved spacers of different sizes in pRb and p107. It is the pocket regions of pRb and p107 that associate with the E2F transcription factor. pRb2 also contains the conserved pocket region suggesting that it has functional similarities to pRb and p107.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rb2
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Chicken, Drosophila, Human, Mouse, Rat
IgG2a
Western Blot
10
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.