Learn More
RACK1 Mouse, Unlabeled, Clone: 20, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610177
Description
Several proteins which specifically bind to PKC have been classified as RACKs (receptors for activated C-kinase). RACK1 was cloned from a rat brain cDNA expression library by screening for proteins that bind PKC in the presence of phosphatidylserine, diacylglycerol, and calcium in a PKC overlay assay. By sequence homology, RACK1 appears to belong to a superfamily that includes the β subunit of G proteins. All of these proteins contain five to eight internal repeat elements known as WD40 motifs, which appear to have a role in protein-protein interactions. In addition, RACK1 contains two short sequences homologous to a PKC-binding sequence identified in Annexin I and in the brain PKC inhibitor KCIP. The binding of RACK1 to PKC is dose-dependent and occurs at a site on PKC that is distinct from the catalytic domain, indicating that RACK1 is not a PKC substrate.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| RACK1 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 50 μg | |
| Cell Biology | |
| Bovine, Canine, Chicken, Frog, Human, Mouse, Rat | |
| IgM |
| Western Blot | |
| 20 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.