missing translation for 'onlineSavingsMsg'
Learn More

Rab11 Mouse, Unlabeled, Clone: 47, BD

Mouse Monoclonal Antibody

£163.84 - £284.36

Specifications

Antigen Rab11
Clone 47
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15825569
View Documents
BD Biosciences
610656
50 μg
ONLINE EXCLUSIVE PRICE
£171.00
£163.84 / 1 set
Save £7.16
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15835569
View Documents
BD Biosciences
610657
150 μg
ONLINE EXCLUSIVE PRICE
£297.00
£284.36 / 1 set
Save £12.64
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Rab proteins are small GTP-binding molecules that are localized to specific intracellular vesicles and organelles. It has been proposed that Rab proteins cycle between GTP- and GDP-bound forms and that this is related to their function as regulators of vesicular traffic. The Rab11 gene encodes a 24 kDa protein of 214 amino acids that has been detected in liver, brain, testis, spleen, and heart. Rab11 protein was isolated from the golgi-microsomal fraction of rat liver and has been detected in the Trans-golgi Network, secretory vesicles, and the pericentriolar recycling endosomes. The distribution of Rab11 indicates that this small protein is involved in regulating traffic at the Golgi complex.

Specifications

Specifications

Rab11
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG2a
Affinity Purified
47
Western Blot
Unconjugated
RUO
Canine, Chicken, Human, Mouse, Rat
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.