missing translation for 'onlineSavingsMsg'
Learn More

Rab11 Mouse, Unlabeled, Clone: 47, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610657

 View more versions of this product

Product Code. 15835569

  • ONLINE EXCLUSIVE PRICE
    £297.00
    £284.36 / 1 set
    Save £12.64
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Rab proteins are small GTP-binding molecules that are localized to specific intracellular vesicles and organelles. It has been proposed that Rab proteins cycle between GTP- and GDP-bound forms and that this is related to their function as regulators of vesicular traffic. The Rab11 gene encodes a 24 kDa protein of 214 amino acids that has been detected in liver, brain, testis, spleen, and heart. Rab11 protein was isolated from the golgi-microsomal fraction of rat liver and has been detected in the Trans-golgi Network, secretory vesicles, and the pericentriolar recycling endosomes. The distribution of Rab11 indicates that this small protein is involved in regulating traffic at the Golgi complex.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rab11
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG2a
Western Blot
47
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.