Learn More
Pax-5 Mouse, Unlabeled, Clone: 24, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610862
Description
There are at least nine members of the paired box (Pax) gene family whose protein products are transcription factors involved in development. The conserved paired box DNA-binding domain is found in the N-terminal region of Pax proteins. An octomer and homeodomain sequence are conserved in the center of the proteins. The Ser/Thr/Pro-rich region in the C-terminal portion contains a conserved 100 amino acid transactivating domain. One of the best studied Pax family members, Pax 5, is a B cell specific activator protein (BSAP). In the early stages of B cell development, Pax-5 influences the expression of several B-cell-specific genes, such as CD19 and CD20. Pax-5 is expressed primarily in pro-, pre-, and mature B cells, but not in plasma cells. Interestingly, Pax-5 mRNA is transiently detected in the mesencephalon and spinal cord during embryogenesis. Expression then shifts to the fetal liver and correlates with the onset of B lymphopoiesis. Pax-5 has been found to be important in both B cell and nervous system development.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
| Pax-5 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Human Pax-5 aa. 151-306 | |
| 50 μg | |
| Cell Biology | |
| Human, Mouse | |
| IgG1 |
| Western Blot | |
| 24 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.