Learn More
p190 Mouse, Unlabeled, Clone: 30, BD
Mouse Monoclonal Antibody
£190.20 - £268.92
Specifications
| Antigen | p190 |
|---|---|
| Clone | 30 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15875289
|
BD Biosciences
610149 |
50 μg |
£198.00 £190.20 / 1 set Save £7.80 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15885289
|
BD Biosciences
610150 |
150 μg |
£280.00 £268.92 / 1 set Save £11.08 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GTPase activating proteins (GAPs) stimulate the GTP-hydrolyzing activity of GTPases, such as p21[ ras]. Ras-GAP (120kDa) is perhaps the best characterized GAP. p190 is a Ras-GAP associated protein that is tyrosine phosphorylated in transformed and growth factor-stimulated cells. Ras-GAP and p190 are targets of oncoproteins and growth factor receptors. Recombinant p190 enhances GTPase activity of the rho family of GTPases. The complete sequence of p190 exhibits three major regions: an N-terminal region with homology to all GTPases, a middle region homologous to transcriptional repressors, and a C-terminal region the is similar to n-chimerin and other small GTPases.
Specifications
| p190 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 30 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Chicken, Frog, Human, Mouse, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.