Learn More
p190 Mouse, Unlabeled, Clone: 30, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610150
Description
GTPase activating proteins (GAPs) stimulate the GTP-hydrolyzing activity of GTPases, such as p21[ ras]. Ras-GAP (120kDa) is perhaps the best characterized GAP. p190 is a Ras-GAP associated protein that is tyrosine phosphorylated in transformed and growth factor-stimulated cells. Ras-GAP and p190 are targets of oncoproteins and growth factor receptors. Recombinant p190 enhances GTPase activity of the rho family of GTPases. The complete sequence of p190 exhibits three major regions: an N-terminal region with homology to all GTPases, a middle region homologous to transcriptional repressors, and a C-terminal region the is similar to n-chimerin and other small GTPases.
Specifications
| p190 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 150 μg | |
| Cell Biology | |
| Canine, Chicken, Frog, Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 30 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.