missing translation for 'onlineSavingsMsg'
Learn More

Human Notch-1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 1 publication

£170.00 - £1585.00

Specifications

Antigen Notch-1
Clone 527425
Concentration LYOPH
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Flow Cytometry, CyTOF, ELISA
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18745209
GREENER_CHOICE
View Documents
R&D Systems
MAB5317-SP
25 μg
£170.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18139940
View Documents
R&D Systems
MAB5317-100
100 μg
£399.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18159270
GREENER_CHOICE
View Documents
R&D Systems
MAB5317-500
500 μg
£1585.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Notch-1 Monoclonal specifically detects Notch-1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

Notch-1
LYOPH
Flow Cytometry, CyTOF, ELISA
Unconjugated
Mouse
Human
P46531
4851
Mouse myeloma cell line NS0-derived recombinant human Notch-1 aa 19-526 Accession # P46531
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Notch-1.
527425
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, Notch1, TAN1, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1
NOTCH1
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.