missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Notch-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB5317-SP
This item is not returnable.
View return policy
Description
Notch-1 Monoclonal specifically detects Notch-1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
| Notch-1 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P46531 | |
| NOTCH1 | |
| Mouse myeloma cell line NS0-derived recombinant human Notch-1 aa 19-526 Accession # P46531 | |
| 25 μg | |
| Primary | |
| Detects human Notch-1. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF, ELISA | |
| 527425 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, Notch1, TAN1, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4851 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction