missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRF-1 Antibody (57-6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
£183.00 - £428.00
Specifications
| Antigen | TRF-1 |
|---|---|
| Clone | 57-6 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 0.5 ug/ml, Simple Western 1:200, Flow Cytometry 1 ug per million cells, ELISA 1:100-1:2000, ICC/IF (Negative) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18217340
|
Novus Biologicals
NB110-68281SS |
0.025 mL |
£183.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18755033
|
Novus Biologicals
NB110-68281 |
0.1 mL |
£428.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRF-1 Monoclonal specifically detects TRF-1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, ICC/IF (Negative).Specifications
| TRF-1 | |
| 1.0 mg/ml | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
| P54274 | |
| 7013 | |
| Recombinant human TRF1 [UniProt# P54274] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 57-6 | |
| Western Blot 0.5 ug/ml, Simple Western 1:200, Flow Cytometry 1 ug per million cells, ELISA 1:100-1:2000, ICC/IF (Negative) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Monkey | |
| NIMA-interacting protein 2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1 | |
| TERF1 | |
| IgG1 κ | |
| Protein G purified | |
| 60 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title