missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRF-1 Antibody (57-6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB110-68281SS
This item is not returnable.
View return policy
Description
TRF-1 Monoclonal specifically detects TRF-1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, ICC/IF (Negative).
Specifications
| TRF-1 | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot 0.5 ug/ml, Simple Western 1:200, Flow Cytometry 1 ug per million cells, ELISA 1:100-1:2000, ICC/IF (Negative) | |
| NIMA-interacting protein 2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1 | |
| Mouse | |
| 60 kDa | |
| 0.025 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
| 7013 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
| 57-6 | |
| Unconjugated | |
| P54274 | |
| TERF1 | |
| Recombinant human TRF1 [UniProt# P54274] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction