missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse CD81 (EAT2), Proteintech
Hamster Monoclonal Antibody
£40.00 - £106.00
Specifications
| Antigen | CD81 |
|---|---|
| Clone | EAT2 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234928
|
Proteintech
65220-1-IG-500UG |
500 μg |
£112.00 £106.00 / 500µg Save £6.00 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236911
|
Proteintech
65220-1-IG-100UG |
100 μg |
£44.00 £40.00 / 100µg Save £4.00 9% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD81 antibody from Proteintech is a armenian hamster monoclonal antibody generated with cd81+ mouse b lymphoma 38c13 of mouse CD81. This antibody recognizes mouse antigen. The CD81 antibody has been validated for the following applications: FC analysis.Specifications
| CD81 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 12520 | |
| IgG | |
| Protein G purification | |
| Flow Antibody |
| EAT2 | |
| Flow Cytometry | |
| Unconjugated | |
| Armenian Hamster | |
| Mouse | |
| CD81+ mouse B lymphoma 38C13 | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title