Learn More
Anti-Mouse CD81 (EAT2), Proteintech
Hamster Monoclonal Antibody
Brand:  Proteintech 65220-1-IG-500UG
 View more versions of this product
Product Code. 30234928
  • ONLINE EXCLUSIVE PRICE
    £112.00
    £106.00 / 500µg
    Save £6.00
    5% Off
Description
The CD81 antibody from Proteintech is a armenian hamster monoclonal antibody generated with cd81+ mouse b lymphoma 38c13 of mouse CD81. This antibody recognizes mouse antigen. The CD81 antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
CD81
Monoclonal
0.5 mg/mL
Armenian Hamster
Protein G purification
RUO
12520
Store at 2-8°C. Stable for one year after shipment.
Liquid
Flow Cytometry
EAT2
Unconjugated
CD81+ mouse B lymphoma 38C13
500 μg
Primary
Mouse
Flow Antibody