missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse CD324 (E-cadherin) (DECMA-1), Proteintech
Rat Monoclonal Antibody
£110.00 - £340.00
Specifications
| Antigen | CD324 (E-cadherin) |
|---|---|
| Clone | DECMA-1 |
| Concentration | 0.5 mg/mL |
| Applications | Immunofluorescence, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237020
|
Proteintech
65241-1-IG-500UG |
500 μg |
£346.00 £340.00 / 500µg Save £6.00 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234399
|
Proteintech
65241-1-IG-100UG |
100 μg |
£110.71 £110.00 / 100µg Save £0.71 0.64% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD324 (E-cadherin) antibody from Proteintech is a rat monoclonal antibody generated with mouse embryonal carcinoma cell line pcc4 aza ri of mouse CD324 (E-cadherin). This antibody recognizes human, mouse, canine antigen. The CD324 (E-cadherin) antibody has been validated for the following applications: IF-P, FC analysis.Specifications
| CD324 (E-cadherin) | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 12550 | |
| IgG1 κ | |
| Affinity purification | |
| Flow Antibody |
| DECMA-1 | |
| Immunofluorescence, Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Human, Mouse, Canine | |
| Mouse embryonal carcinoma cell line PCC4 Aza RI | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title