missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse CD324 (E-cadherin) (DECMA-1), Proteintech
Rat Monoclonal Antibody
Brand: Proteintech 65241-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD324 (E-cadherin) antibody from Proteintech is a rat monoclonal antibody generated with mouse embryonal carcinoma cell line pcc4 aza ri of mouse CD324 (E-cadherin). This antibody recognizes human, mouse, canine antigen. The CD324 (E-cadherin) antibody has been validated for the following applications: IF-P, FC analysis.Specifications
| CD324 (E-cadherin) | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Affinity purification | |
| RUO | |
| 12550 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence, Flow Cytometry | |
| DECMA-1 | |
| Unconjugated | |
| Mouse embryonal carcinoma cell line PCC4 Aza RI | |
| 500 μg | |
| Primary | |
| Human, Mouse, Canine | |
| Flow Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction