Learn More
Moesin Mouse, Unlabeled, Clone: 38, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610401
Additional Details : Weight : 0.01000kg
Description
Moesin belongs to a family of proteins that includes ezrin and radixin. These proteins are co-expressed in a variety of cells. They localize to the cytoskeleton and modify interactions between cytoskeletal and membrane proteins. There is approximately 70% homology between moesin and ezrin and 80% homology between moesin and radixin. Moesin is intracellular and contains multiple conserved domains that are putative phosphorylation sites. It has a calculated molecular weight of 67.8 kDa, but migrates in SDS-PAGE at approximately 78 kDa. This discrepancy is likely due to a charged alpha-helical region within the protein.
Specifications
Moesin | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
50 μg | |
Cell Biology | |
Canine, Chicken, Human, Mouse, Rat | |
IgG1 |
Western Blot | |
38 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.