Learn More
Moesin Mouse, Unlabeled, Clone: 38, BD
Mouse Monoclonal Antibody
£237.00 - £428.00
Specifications
Antigen | Moesin |
---|---|
Clone | 38 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15865439
|
BD Biosciences
610402 |
150 μg |
£428.00
1 set |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15855439
|
BD Biosciences
610401 |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Moesin belongs to a family of proteins that includes ezrin and radixin. These proteins are co-expressed in a variety of cells. They localize to the cytoskeleton and modify interactions between cytoskeletal and membrane proteins. There is approximately 70% homology between moesin and ezrin and 80% homology between moesin and radixin. Moesin is intracellular and contains multiple conserved domains that are putative phosphorylation sites. It has a calculated molecular weight of 67.8 kDa, but migrates in SDS-PAGE at approximately 78 kDa. This discrepancy is likely due to a charged alpha-helical region within the protein.
Specifications
Moesin | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
38 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.